Learn More
Karl Hecht™ Water Bath Rack, Plexiglass™
Product Code. 10257661
Change view
Click to view available options
Diameter (Metric) Min. Opening:
16 x 23mm diameter holes
20 x 18mm diameter holes
24 x 11mm diameter holes
24 x 12mm diameter holes
Holds:
16 small Boehringer/Merck bottles
20 Centrifuge Tubes, Test Tubes
24 1.5 mL Reaction Tubes
24 Coagulometer Tubes
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Min. Opening Holds unitSize
10257661 24 x 12mm diameter holes 24 Coagulometer Tubes Each
10585002 16 x 23mm diameter holes 16 small Boehringer/Merck bottles Each
10595002 20 x 18mm diameter holes 20 Centrifuge Tubes, Test Tubes Each
10630592 24 x 11mm diameter holes 24 1.5 mL Reaction Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10257661 Supplier Karl Hecht™ Supplier No. 43188006
Only null left
Add to basket
Add to basket
Perfect for hanging into a water bath tank. Karl Hecht™ Water Bath Rack, Plexiglass™ comes in a variety of size and array configurations.
  • Durable Plexiglass™ construction.
  • Hanging handles.
  • Available in a variety of size and array configurations.
Specifications
For Use With (Equipment) Water Bath
Holds 24 Coagulometer Tubes
Material Plexiglass™
Diameter (Metric) Min. Opening 24 x 12mm diameter holes