Learn More
behr Labor-Technik™ behrotest™ Water Sample Homogenizer
Product Code. 10034561
Click to view available options
Volume (Metric):
15 L
25 L
Unit Size:
Each
Product Code. 10034561
Brand: behr Labor-Technik™ B00218341
Only null left
Add to basket
Add to basket
Homogenizing device for the pretreatment, separation and homogenization of heterogeneous water samples in compliance with the standards of the Working Group of the Federal States for Quality Assurance and Control for Waste Water Examinations.
  • Can be carried by two handles
    • Is protected in the in top container against damage
      • Easy filling with flattened drain insert.
        • Scales for exact measurement of sample quantities
          • Directly ready for use by inserting the transport rack on the magnetic stirrer
            • The speed of the magnetic stirrer can be adjusted by means of a regulator.
Specifications
Certifications/Compliance DIN 38402 A 30
Description QMR 25 Homogenization device
Volume (Metric) 25 L
Type Homogenization device