Learn More
Poulten & Graf GmbH Washing Syringe
Brand: Poulten & Graf GmbH 7 30053
Additional Details : Weight : 0.10000kg
Description
- With steel piston
- With 2 tips and metal holding bow
- Interchangeable
- Amber graduation
- For technical use only
Specifications
Autoclavable | |
Clear | |
Yes | |
No | |
1 | |
Yes |
Neutral Glass, Neutral Glass (Barrel), Nickel-plated Brass (Tip), Steel (Plunger) | |
Washing syringe | |
Washing Syringe | |
Amber | |
200 mL |