Learn More
Robu Washing-Bottle lower part 250ml, NS 45/40
Brand:  Robu 34270
Washing-Bottle lower part 250ml, NS 45/40
Product Code. 10012091
  • ONLINE EXCLUSIVE PRICE
    77.80 €
    68.65 € / Each
    Save 9.15 €
    11% Off
Only null left
Estimated Shipment: 10-02-2026