Learn More
Smeg™ Washer Rack
Product Code. 11925574
Click to view available options
Rack Type:
Bottom rack with hole for positioningon the top level
Complete bottom rack for positioning on the bottom level only
Half bottom rack for positioning on the top level only
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 11925574
Brand: Smeg™ PF1
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Designed for use with Smeg™ glassware washing devices. Smeg Washer Racks can be used as racks to hold small glass items or as an alternative to the Nylon holding nets.
  • Additional accessories for the small glassware baskets CS1-1 and CS2
  • Flat AISI 304 stainless steel mesh with small mesh openings 
  • PF1 bottom rack with hole for positioning on the top level
  • PF2 complete bottom rack for positioning on the bottom level only
  • PF1/2 half bottom rack for positioning on the top level only
Specifications
Material Stainless Steel
Product Type Rack
Rack Type Bottom rack with hole for positioningon the top level
For Use With (Equipment) Small glassware, baskets and holding nets