Learn More
Heathrow Scientific Wash Bottles Safety Labeled, Self Venting, Wide Mouth, 500 mL
Shop All Thermo Scientific CO₂ Incubators ProductsDescription
The Wash Bottle has clear and informative safety information and is specific to the product contained and includes the following — the Chemical Abstract Service (CAS) No., National Fire Protection Association (NFPA) codes and symbols, Protective Equipment Symbol (PES), Target Organ Information and Hazard Codes. The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
- Wide bottle mouth makes filling easier
- Non-metallic vent that prevents the risk of solvent leakage
- Bottle is made from LDPE
- Bottle must only be used with chemicals that are safe and compatible with LDPE
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16.9 oz. |
| Color | Clear |
| Description | Wash Bottle Safety Labeled |
| For Use With (Application) | Chemical |
| Material | LDPE |
| No. per Case | 24/Cs. |
| No. per Pack | 6/Pk. |
| Packaging | Box |
| Sterility | Non-sterile |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.