missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific Wash Bottles Safety Labeled, Self Venting, Wide Mouth, 500 mL

Product Code. 16300726 Shop All Thermo Scientific CO₂ Incubators Products
Click to view available options
Chemical Label:
Acetone
Assorted
Blank
Distilled Water
Ethanol
Isopropanol
Methanol
Sodium Hypochlorite
Closure Color:
Assorted
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 16300726

Brand: Heathrow Scientific HS120253

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Wash Bottle featuring a closure and stem that are molded in one piece, making them leak-proof.

The Wash Bottle has clear and informative safety information and is specific to the product contained and includes the following — the Chemical Abstract Service (CAS) No., National Fire Protection Association (NFPA) codes and symbols, Protective Equipment Symbol (PES), Target Organ Information and Hazard Codes. The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.

  • Wide bottle mouth makes filling easier
  • Non-metallic vent that prevents the risk of solvent leakage
  • Bottle is made from LDPE
  • Bottle must only be used with chemicals that are safe and compatible with LDPE

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Color Transparent
Description Wash Bottle Safety Labeled
For Use With (Application) Chemical
Material LDPE
No. per Case 24/Cs.
No. per Pack 6/Pk.
Packaging Box
Sterility Non-sterile
Width (Metric) 7.00 cm
Closure Color White
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 22.86 cm
Product Type Wash Bottle (Ethanol)
Width (English) 2.75 in.
Height (English) 9 in.
Chemical Label Ethanol
Closure Material Polypropylene
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.