missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Wash Bottles, Transparent Polypropylene
Description
Wash bottles are appropriate for cleaning, coating and dousing in any laboratory environment. Polypropylene bottles are ideal for a variety of applications, including use with a range of chemicals, solvents and temperatures. Spray bottles are individually wrapped in clearly labeled polyethylene bags.
- Designed for precise spray control
- Finely drawn tip creates a precise spray jet for fine control
- Optimised medium backflow through the continuous spray tube
- Choice of narrow- or wide-mouth bottles
- Narrow-mouth, tapered bottles for easier handling
- Wide-mouth, square-shouldered bottles for easy and safe filling
- All bottles feature screw cap lids with GL threads for a strong, secure hold
- Constructed of durable, transparent polypropylene
- Chemically resistant for use with liquid or powdered reagents
- Temperature resistant and autoclavable
- Transparent material for easy identification and monitoring
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Transparent |
| Material | Polypropylene |
| Quantity | 12Pack |
| Thread Size (Metric) | GL 25 |
| Mouth | Narrow |
| Height (Metric) | 180 mm |
| Product Type | Wash Bottle |
| Diameter (Metric) | 74 mm |
| Closure Type | Screw Cap |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?