Learn More
Brannan™ Wall Thermometer with Calibration Certificate
Product Code. p-200007080
Click to view available options
Thermometer Fill:
Blue Spirit
Red Spirit Kerosene
Unit Size:
Each
Product Code. 17843088
Brand: Brannan™ 14/435/3 + 49/101/0
Only null left
Add to basket
Add to basket
Wall thermometer with clear black printed temperature scales. Complete with traceable calibration.
  • Highly visible wall thermometer
  • Easy to read temperature scales in Celsius and Fahrenheit
  • Supplied with calibration
  • Temperature range -20 to 50°C
  • Suitable for indoor and outdoor use
  • Made in UK
Specifications
Calibration Points 0°C, 20°C, 35°C
Certifications/Compliance UK National Standards ISO 9001
Height (Metric) 21.5 cm
Length (Metric) 0.8 cm
Material Plastic
Thermometer Fill Red Spirit Kerosene
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) -4°F
Min. Temperature (Metric) -20°C
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Accuracy ± 1°C at 10°C
Subdivision 0.1°C
Width (Metric) 4 cm