Learn More
Brannan™ Spirit-filled Max Min Thermometer
Product Code. 13029212
Click to view available options
Temperature Range (Metric):
-30°C to +50°C
Unit Size:
Each
Product Code. 13029212
Brand: Brannan™ 63/096/0
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Measures maximum and minimum temperaturess with non-toxic spirit filling. Brannan™ Spirit-filled Max Min Thermometer can be wall-mounted and resets with the push of a button.
Measures maximum and minimum temperatures with non-toxic spirit filling.
  • Wall mounting in plastic case
  • 205mm long.
  • Push button reset.
Specifications
Length (Metric) 205 mm
Thermometer Fill Non-toxic Spirit
Max. Temperature (Metric) 50°C
Min. Temperature (Metric) -30°C
Temperature Range (Metric) -30°C to +50°C