missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VX 765

Product Code. 18787576
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18787576 10 mg 10mg
18717036 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18787576 Supplier Tocris Bioscience™ Supplier No. 7143/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity and selective caspase-4 and caspase 1/ICE inhibitor; orally bioavailable

VX 765 is a high affinity and selective caspase-4 and caspase 1/ICE inhibitor (Ki of <0.6 and 0.8 nM, respectively). Prodrug of VRT-043198. Inhibits LPS-induced IL-1β production ∼ 60% in collagen-induced arthritis mouse model. Exhibits anticonvulsant effects in mouse model of acute seizures. Anti-inflammatory. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4-Amino-3-chlorobenzoyl)-3-methyl-L-valyl-N-[(2 R,3 S)-2-ethoxytetrahydro-5-oxo-3-furanyl]-L-prolinamide
CAS 273404-37-8
Quantity 10 mg
Target Caspase Inhibitors
Molecular Formula C24H33ClN4O6
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.