Learn More
VUF10460, MedChemExpress
Product Code. 16406087
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16406087
Brand: Medchem Express HY101420
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress VUF10460 is a non-imidazole histamine H4 receptor agonist; binds to rat H4 receptor with a pKi of 7.46.
VUF10460 is a non-imidazole histamine H4 receptor agonist
Specifications
Chemical Name or Material VUF10460
CAS 1028327-66-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H19N5
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (185.64 mM; Need ultrasonic)
SMILES NC1=NC(C2=CC=CC=C2)=CC(N3CCN(C)CC3)=N1
Molecular Weight (g/mol) 269.34
Formula Weight 269.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation