Learn More
VU0650786, MedChemExpress
Product Code. 16423006
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423006 25 mg 25mg
16416957 5 mg 5mg
16404238 10 mg 10mg
16400577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16423006 Supplier Medchem Express Supplier No. HY108710
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress VU0650786 is a potent and selective CNS penetrant negative allosteric modulator of metabotropic glutamate receptor subtype 3 (mGlu3 NAM), with an IC50 of 392 nM. VU0650786 has antidepressant and anxiolytic activity in rodents.
VU0650786 is a potent and selective CNS penetrant negative allosteric modulator of metabotropic glutamate receptor subtype 3 (mGlu3 NAM), with an IC50 of 392 nM. VU0650786 has antidepressant and anxiolytic activity in rodents.
Specifications
Chemical Name or Material VU0650786
CAS 1809085-30-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H15ClFN5O2
Quantity 25 mg
Solubility Information DMSO : 250 mg/mL (644.66 mM; Need ultrasonic)
SMILES ClC1=CN=C(OCC2=NN([C@H](C)CN(C3=C(F)N=CC=C3)C4=O)C4=C2)C=C1
Molecular Weight (g/mol) 387.8
Formula Weight 387.8
Percent Purity 99.16%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation