missing translation for 'onlineSavingsMsg'
Learn More

VU0650786, MedChemExpress

Product Code. 16400577
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16400577 50 mg 50mg
16423006 25 mg 25mg
16416957 5 mg 5mg
16404238 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16400577 Supplier Medchem Express Supplier No. HY108710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VU0650786 is a potent and selective CNS penetrant negative allosteric modulator of metabotropic glutamate receptor subtype 3 (mGlu3 NAM), with an IC50 of 392 nM. VU0650786 has antidepressant and anxiolytic activity in rodents.

VU0650786 is a potent and selective CNS penetrant negative allosteric modulator of metabotropic glutamate receptor subtype 3 (mGlu3 NAM), with an IC50 of 392 nM. VU0650786 has antidepressant and anxiolytic activity in rodents.

Specifications

Chemical Name or Material VU0650786
CAS 1809085-30-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H15ClFN5O2
Quantity 50 mg
Solubility Information DMSO : 250 mg/mL (644.66 mM; Need ultrasonic)
SMILES ClC1=CN=C(OCC2=NN([C@H](C)CN(C3=C(F)N=CC=C3)C4=O)C4=C2)C=C1
Molecular Weight (g/mol) 387.8
Formula Weight 387.8
Percent Purity 99.16%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.