Learn More
VU0364770, MedChemExpress
Product Code. 16484386
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16484386
Brand: Medchem Express HY100588
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress VU0364770 is a selective and potent positive allosteric modulator (PAM) of mGlu4.
VU0346770 exhibits EC50s of 290 nM and 1.1 μM at rat mGlu4 and human mGlu4 receptor, respectively. VU0364770 exhibits antagonist activity at mGlu5 with a potency of 17.9 μM and PAM activity at mGlu6 with a potency of 6.8 μM. VU0364770 also possesses activity at MAO with Ki values of 8.5 and 0.72 μM for human MAO-A and human MAO-B, respectively.
Specifications
Chemical Name or Material VU0364770
CAS 61350-00-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C12H9ClN2O
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (429.79 mM)
SMILES O=C(C1=NC=CC=C1)NC2=CC=CC(Cl)=C2
Molecular Weight (g/mol) 232.67
Formula Weight 232.67
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation