Learn More
VU0361737, MedChemExpress
Product Code. 16486456
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486456 100 mg 100mg
16466058 10 mg 10mg
16452197 50 mg 50mg
16428667 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16486456 Supplier Medchem Express Supplier No. HY14418
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress VU0361737 (ML-128) is a potent, selective and CNS penetrant positive allosteric modulator of metabotropic glutamate receptor 4 (mGluR4 PAM), with EC50s of 240 nM and 110 nM for human and rat mGluR4 receptors, respectively. VU0361737 has neuroprotective effect. VU0361737 is potential for Parkinson's disease research.
VU0361737 (ML-128) is a potent, selective and CNS penetrant positive allosteric modulator of metabotropic glutamate receptor 4 (mGluR4 PAM), with EC50s of 240 nM and 110 nM for human and rat mGluR4 receptors, respectively. VU0361737 has neuroprotective effect. VU0361737 is potential for Parkinson's disease research.
Specifications
Chemical Name or Material VU0361737
CAS 1161205-04-4
Color White
Purity Grade Notes Research
Molecular Formula C13H11ClN2O2
Quantity 100 mg
Synonym ML-128
Solubility Information DMSO : 100 mg/mL (380.68 mM; Need ultrasonic)
SMILES ClC(C=C1)=C(OC)C=C1NC(C2=NC=CC=C2)=O
Molecular Weight (g/mol) 262.69
Formula Weight 262.69
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation