missing translation for 'onlineSavingsMsg'
Learn More

VU0357017 hydrochloride, MedChemExpress

Product Code. 16480348
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480348

Brand: Medchem Express HY19752A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VU0357017 hydrochloride (CID-25010775) is a potent, selective and brain-penetrant allosteric agonist of M1 muscarinic acetylcholine receptor, with an EC50 of 477 nM. VU0357017 hydrochloride is highly selective for M1 and has no activity at M2-M5 up to the highest concentrations tested (30 μM). VU0357017 hydrochloride can be used for the research of Alzheimer’s disease and schizophrenia.

VU0357017 hydrochloride (CID-25010775) is a potent, selective and brain-penetrant allosteric agonist of M1 muscarinic acetylcholine receptor, with an EC50 of 477 nM. VU0357017 hydrochloride is highly selective for M1 and has no activity at M2-M5 up to the highest concentrations tested (30 μM). VU0357017 hydrochloride can be used for the research of Alzheimer’s disease and schizophrenia.

Specifications

Chemical Name or Material VU0357017 hydrochloride
CAS 1135242-13-5
Color White
Purity Grade Notes Research
Molecular Formula C18H28ClN3O3
Quantity 5 mg
Synonym CID-25010775
Solubility Information H2O : 33.33 mg/mL (90.11 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (67.59 mM; Need ultrasonic)
SMILES O=C(N1CCC(NCCNC(C2=CC=CC=C2C)=O)CC1)OCC.[H]Cl
Molecular Weight (g/mol) 369.89
Formula Weight 369.89
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.