Learn More
Tocris Bioscience™ VU 0542270
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
VU 0542270 is a selective inhibitor of Kir6.1/SUR1 KATP channels (IC50 = 104 nM). Shows no activity at other Kir channel types at up to 30 μM. Dose-dependently induces constriction of ductus arteriosus vessels.
Specifications
Specifications
| Chemical Name or Material | 1-[(4-Methyl-2-thiophen-2-yl-1,3-thiazol-5-yl)methyl]-3-thiophen-2-ylurea |
| CAS | 1396814-79-1 |
| Quantity | 50 mg |
| Target | Inward Rectifier Potassium Channel Activators |
| Molecular Formula | C14H13N3OS3 |
| Purity | 0.98 |