Learn More
VU 0238429, MedChemExpress
Product Code. 16497727
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16497727
Brand: Medchem Express HY12157
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress VU 0238429 is positive allosteric modulator of muscarinic acetylcholine receptor subtype 5 (mAChR5 or M5), with an EC50 of 1.16 μM.
VU 0238429 is positive allosteric modulator of muscarinic acetylcholine receptor subtype 5 (mAChR5 or M5), with an EC50 of 1.16 μM.
Specifications
Chemical Name or Material VU 0238429
CAS 1160247-92-6
Color Orange
Purity Grade Notes Research
Molecular Formula C17H12F3NO4
Quantity 5 mg
Solubility Information DMSO : ≥ 150 mg/mL (427.01 mM)
SMILES O=C1N(CC2=CC=C(OC)C=C2)C3=C(C=C(OC(F)(F)F)C=C3)C1=O
Molecular Weight (g/mol) 351.28
Formula Weight 351.28
Percent Purity 99.96%
Grade Research
Physical Form Solid