Learn More
VTP-27999 Hydrochloride, MedChemExpress
Product Code. 16438218
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16438218
Brand: Medchem Express HY76652
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress VTP-27999 Hcl is an alkyl amine Renin inhibitor; VTP-27999 is useful for Hypertension and End-Organ Diseases.
VTP-27999 Hcl is an alkyl amine Renin inhibitor
Specifications
Chemical Name or Material VTP-27999 Hydrochloride
Physical Form Solid
CAS 1264191-73-2
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C26H42Cl2N4O5
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : ≥ 100 mg/mL (178.08 mM)
SMILES O=C(NC[C@H](C[C@@H]1COCCC1)NC)N(CCC2)C[C@]2([H])[C@](OCCNC(OC)=O)([H])C3=CC(Cl)=CC=C3.Cl
Molecular Weight (g/mol) 561.54
Formula Weight 561.54
Percent Purity 98.02%
Grade Research