missing translation for 'onlineSavingsMsg'
Learn More

VTI1B Antibody [HRP], Novus Biologicals Biologicals™

Product Code. 30500448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30500448 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30500448 Supplier Novus Biologicals Supplier No. NBP327901H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VTI1B Polyclonal antibody specifically detects VTI1B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VTI1B
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias vesicle transport through interaction with t-SNAREs homolog 1B (yeast), Vesicle transport v-SNARE protein Vti1-like 1, vesicle-associated soluble NSF attachment protein receptor, v-SNARE, VTI1, VTI1L, VTI1L1, VTI1-LIKE, Vti1-rp1, VTI2vesicle transport through interaction with t-SNAREs homolog 1B
Host Species Rabbit
Immunogen VTI1B antibody was raised against a 17 amino acid peptide near the center of human VTI1B. The immunogen is located within amino acids 150 - 200 of VTI1B.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10490
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.