missing translation for 'onlineSavingsMsg'
Learn More

VTCN1 Mouse anti-Human, Clone: 2E5B1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66817-1-IG-20UL

 View more versions of this product

Product Code. 16829045

  • 145.00 € / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

B7-H4 (B7x, VTCN1) a costimulatory protein which is reported to function as a negative regulator of T-cell mediated immunity. Although B7-H4 binds an unknown receptor, it is thought to deliver an inhibitory signal to T-cells preventing their proliferation, cell cycle progression and interleukin-2 production. B7-H4 deficient mice are only minimally affected; suggesting B7-H4 is important in the fine tuning of the T-cell mediated immune response. B7-H4 is expressed on activated T-cells, B-cells, monocytes and dendritic cells. Aberrant expression has been associated with cancers of the lung, breast and ovary in humans. B7-H4 is expressed primarily on the membrane of lymphoid cells, and as an immunoinhibitory protein that interacts with receptors on the surface of T lymphocytes, it is involved in mediating cellular and humoral immune responses. Overexpression of B7-H4 is associated with certain maligcies, including ovarian and breast cancer, which may be a mechanism by which tumor cells suppress T cell immunity and facilitate tumor progression. Further, VTCN1 may be a useful biomarker for the early detection of ovarian cancer and Renal Cell Carcinoma (RCC).
Specifications

Specifications

VTCN1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q7Z7D3
VTCN1
VTCN1 Fusion Protein Ag27751
20 μL
Primary
Human
Antibody
IgG1
Western Blot
2E5B1
Unconjugated
VTCN1
B7 H4, B7h.5, B7H4, B7S1, B7X, FLJ22418, PRO1291, Protein B7S1, RP11 229A19.4, VCTN1, VTCN1
Mouse
Protein G
RUO
79679
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.