missing translation for 'onlineSavingsMsg'
Learn More

VS-5584, MedChemExpress

Product Code. 16419637
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16419637

Brand: Medchem Express HY16585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VS-5584 is a pan-PI3K/mTOR kinase inhibitor with IC50s of 16 nM, 68 nM, 42 nM, 25 nM, and 37 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR, respectively. VS-5584 simultaneously blocks mTORC2 as well as mTORC1.

VS-5584 is a pan-PI3K/mTOR kinase inhibitor with IC50s of 16 nM, 68 nM, 42 nM, 25 nM, and 37 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR, respectively. VS-5584 simultaneously blocks mTORC2 as well as mTORC1.

Specifications

Chemical Name or Material VS-5584
CAS 1246560-33-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H22N8O
Quantity 5 mg
Synonym SB2343
Solubility Information DMSO : 33.33 mg/mL (94.04 mM; Need ultrasonic)
SMILES NC1=NC=C(C2=C3N=C(C)N(C(C)C)C3=NC(N4CCOCC4)=N2)C=N1
Molecular Weight (g/mol) 354.41
Formula Weight 354.41
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.