Learn More
Grant Instruments™ VR Rack for Grant Heated Circulating Bath
Description
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
- Accommodates tubes as small as 0.5mL or as large as 30mm dia.
- For use with Grant Heated Circulating Baths and Heating Circulators.
Specifications
Specifications
| Capacity (Metric) | 0.5 mL |
| For Use With (Equipment) | T100, TC120, TX150, TXF200 Heating Circulators |
| Holds | (14) 30 mm Tubes |