missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VPS26B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15756161 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15756161 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15756161 Supplier Invitrogen™ Supplier No. PA558282

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RRYFKQQEVV LWRKGDIVRK SMSHQAAIAS QRFEGTTSLG EVRTPSQLSD NNCRQ Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

In mammals, there are two paralogues of yeast Vps26, VPS26A and VPS26B. VPS26 is a component of the retromer complex composed of VPS26 (VPS26A or VPS26B), VPS29, VPS35, SNX1 and SNX2. VPS26A and VPS26B subunits define distinct retromer complexes. The retromer complex is important in recycling transmembrane receptors from endosomes to the trans-Golgi network (TGN).
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS26B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS26B
Gene Accession No. Q4G0F5
Gene Alias 1810012I05Rik; 2310075A12Rik; AI848392; Pep8b; vacuolar protein sorting 26 homolog B (S. pombe); vacuolar protein sorting 26 homolog B (yeast); vacuolar protein sorting-associated protein 26B; Vesicle protein sorting 26B; VPS26 retromer complex component B; VPS26, retromer complex component B; Vps26b
Gene Symbols VPS26B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS26B. Recombinant protein control fragment (Product #RP-97414).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112936
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.