missing translation for 'onlineSavingsMsg'
Learn More

VPS11 Antibody (SR1048), Novus Biologicals™

Product Code. 18640194 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18640194 100 μg 100µL
18669913 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18640194 Supplier Novus Biologicals Supplier No. NBP322131100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

VPS11 Monoclonal antibody specifically detects VPS11 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS11
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone SR1048
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:80, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias hVPS11, PEP5, RING finger protein 108, RNF108END1, vacuolar protein sorting 11 (yeast homolog), vacuolar protein sorting 11 homolog (S. cerevisiae), vacuolar protein sorting protein 11, vacuolar protein sorting-associated protein 11 homolog
Host Species Rabbit
Immunogen A synthesized peptide derived from human VPS11 (Uniprot #: Q9H270)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 55823
Target Species Human, Mouse
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.