missing translation for 'onlineSavingsMsg'
Learn More

Vorinostat, MedChemExpress

Product Code. 16446457
Encompass_Preferred
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Unit Size:
1g
250mg
500mg
5g
This item is not returnable. View return policy

Product Code. 16446457

Brand: Medchem Express HY10221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vorinostat (SAHA) is a potent and orally active pan-inhibitor of HDAC1, HDAC2 and HDAC3 (Class I), HDAC6 and HDAC7 (Class II) and HDAC11 (Class IV), with ID50 values of 10 nM and 20 nM for HDAC1 and HDAC3, respectively. Vorinostat induces cell apoptosis. Vorinostat is also an effective inhibitor of human papillomaviruse (HPV)-18 DNA amplification.

Vorinostat (SAHA) is a potent and orally active pan-inhibitor of HDAC1, HDAC2 and HDAC3 (Class I), HDAC6 and HDAC7 (Class II) and HDAC11 (Class IV), with ID50 values of 10 nM and 20 nM for HDAC1 and HDAC3, respectively. Vorinostat induces cell apoptosis. Vorinostat is also an effective inhibitor of human papillomaviruse (HPV)-18 DNA amplification.

Specifications

Chemical Name or Material Vorinostat
Physical Form Solid
CAS 149647-78-9
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C14H20N2O3
For Use With (Application) Cancer-programmed cell death
Synonym SAHA Suberoylanilide hydroxamic acid
Solubility Information DMSO : ≥ 100 mg/mL (378.33 mM)
SMILES O=C(NC1=CC=CC=C1)CCCCCCC(NO)=O
Molecular Weight (g/mol) 264.32
Formula Weight 264.32
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.