Learn More
Vorapaxar, MedChemExpress
Product Code. p-200099926
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16476017
Brand: Medchem Express HY10119
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Vorapaxar (SCH 530348), an antiplatelet agent, is a selective, orally active, and competitive thrombin receptor protease-activated receptor (PAR-1) antagonist (Ki=8.1 nM). Vorapaxar (SCH 530348) inhibits thrombin receptor-activating peptide (TRAP)-induced platelet aggregation in a dose-dependent manner.
Vorapaxar (SCH 530348), an antiplatelet agent, is a selective, orally active, and competitive thrombin receptor protease-activated receptor (PAR-1) antagonist (Ki=8.1 nM). Vorapaxar (SCH 530348) inhibits thrombin receptor-activating peptide (TRAP)-induced platelet aggregation in a dose-dependent manner.
Specifications
Chemical Name or Material Vorapaxar
CAS 618385-01-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H33FN2O4
Quantity 5 mg
Synonym SCH 530348
Solubility Information DMSO : 25 mg/mL (50.75 mM; Need ultrasonic)
SMILES O=C(O[C@@H]1C)[C@@]2([H])[C@]1([H])[C@@H](/C=C/C3=NC=C(C=C3)C4=CC=CC(F)=C4)[C@]([C@](C5)([H])C2)([H])CC[C@H]5NC(OCC)=O
Molecular Weight (g/mol) 492.58
Formula Weight 492.58
Percent Purity 98.72%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation