Learn More
Karl Hecht™ Volumetric Flask, Conformity Certified, Duran™
Product Code. 10023851
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
1000 mL
2000 mL
5 L
10 L
Closure Size:
NS 10
NS 12
NS 14
NS 24
NS 29
NS 34
NS 45
NS 7
Neck Style:
Standard
Wide
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 10023851
Brand: Karl Hecht™ 42097099
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask, Conformity Certified, Duran™ is thick-walled, clearly marked and labeled, and has a solid finish.
  • Duran™ borosilicate glass 3.3.
  • Fused blue graduation.
  • Standard ground without stopper.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Certifications/Compliance DIN EN ISO 1042
Capacity (Metric) 10 L
Material Borosilicate Glass (3.3)
Neck Style Standard
Product Type Volumetric Flask
Graduated Yes
Closure Size NS 45