Learn More
Poulten & Graf GmbH VOLAC Nessler Tube
Product Code. 10600703
Change view
Click to view available options
:
Capacity, Metric: 50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10600703 Capacity, Metric: 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10600703 Supplier Poulten & Graf GmbH Supplier No. R336/2
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Poulten & Graf GmbH VOLAC Nessler Tube are designed to be as free from color as possible and are ideal for color comparison.
  • Clear glass
  • Formulated to be a free of color as possible
  • Ideal for color comparison
Specifications
Autoclavable Autoclavable
Certifications/Compliance BS 612
Material Glass
Sterility Non-sterile
Quantity 2
Volume (Metric) 50 mL
Type Nessler Tube