Learn More
Poulten & Graf GmbH VOLAC Nessler Tube
Description
- Clear glass
- Formulated to be a free of color as possible
- Ideal for color comparison
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | BS 612 |
| Material | Glass |
| Sterility | Non-sterile |
| Quantity | 2 |
| Volume (Metric) | 50 mL |
| Type | Nessler Tube |