Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations
Product Code. 10260002
Click to view available options
Volume (Metric):
3 mL
4 mL
5 mL
10 mL
15 mL
25 mL
50 mL
100 mL
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy
Product Code. 10260002
Brand: Poulten & Graf GmbH 1 2323902
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations allow accurate measurements of a volume of solution.
  • With 2 marks
  • Blue graduations
  • Conformity approved
  • With batch certificate
Specifications
Certifications/Compliance EN ISO 648
Class AS
Material Soda Lime Glass
Sterility Non-sterile
Graduated Yes
Graduation Color Blue
Graduation Type Ring marks
Quantity 5
Tolerance 0.03 mL
Volume (Metric) 15 mL
Type Two-mark Bulb Pipette
Disposable Reusable