missing translation for 'onlineSavingsMsg'
Learn More

VMD2 293T Cell Overexpression Lysate (Denatured), Abnova

Product Code. 16147475
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16147475 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16147475 Supplier Abnova Supplier No. H00007439T01.100uL
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Western Blotting

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description VMD2 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. BC041664.1
Molecular Weight (g/mol) 66.55
pH Range 6.8
Plasmid pCMV-VMD2 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-VMD2 antibody (H00007439-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.