Learn More
VLX600, TRC
Product Code. 30451453
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451453 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30451453 Supplier Toronto Research Chemicals Supplier No. TRCV7571751MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(2-Pyridinyl)ethanone 2-(6-Methyl-5H-1,2,4-triazino[5,6-b]indol-3-yl)hydrazone
CAS 327031-55-0
Molecular Formula C17H15N7
Quantity 1 mg
Synonym 1-(2-Pyridinyl)ethanone 2-(6-Methyl-5H-1,2,4-triazino[5,6-b]indol-3-yl)hydrazone, VLX600
InChI Formula InChI=1S/C17H15N7/c1-10-6-5-7-12-14(10)19-16-15(12)22-24-17(20-16)23-21-11(2)13-8-3-4-9-18-13/h3-9H,1-2H3,(H2,19,20,23,24)/b21-11+
SMILES C\C(=N/Nc1nnc2c3cccc(C)c3[nH]c2n1)\c4ccccn4
IUPAC Name 6-methyl-N-[(E)-1-pyridin-2-ylethylideneamino]-5H-[1,2,4]triazino[5,6-b]indol-3-amine
Molecular Weight (g/mol) 317.35
Formula Weight 317.1389