Learn More
VL285, MedChemExpress
Product Code. 16477227
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16477227
Brand: Medchem Express HY111663
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress VL285 is a potent VHL ligand with an IC50 of 0.34 μM.
VL285 is a potent VHL ligand with an IC50 of 0.34 μM.
Specifications
Chemical Name or Material VL285
CAS 1448188-57-5
Color White
Purity Grade Notes Research
Molecular Formula C29H32N4O4S
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (234.68 mM; Need ultrasonic)
SMILES CC(N=CS1)=C1C2=CC=C(CNC([C@@H]3C[C@@H](O)CN3C([C@@H](N4CC(C=CC=C5)=C5C4=O)C(C)C)=O)=O)C=C2
Molecular Weight (g/mol) 532.65
Formula Weight 532.65
Percent Purity 98.84%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death