missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Vitronectin XF™

Product Code. 15417440
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417440 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15417440 Supplier STEMCELL Technologies Supplier No. 07180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Matrix for maintenance of human ES/iPS cells in combination with TeSR™-E8™, mTeSR™1, or TeSR™2

  • Vitronectin XF™, developed and manufactured by Nucleus Biologics, is an effective alternative to Corning™ Matrigel™
  • Vitronectin XF™ is a defined, xeno-free cell culture matrix that supports the growth and differentiation of human pluripotent stem cells
  • When used with mTeSR™1, mTeSR™ Plus, TeSR™-E8™, or TeSR™2, Vitronectin XF™ provides a defined culture system for the maintenance of human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells under feeder-free conditions
  • This system allows complete control over the culture environment, resulting in more consistent cell populations and reproducible results in downstream applications
  • Note: Catalog #07180 is the matrix only and requires additional reagents (Catalog #07183, 07174 or 05872, and 27147) available for purchase separately or in a kit with Vitronectin XF™ (Catalog #07190 or 07191)

Specifications

Cell Type Endoderm, PSC-Derived, Pluripotent Stem Cells
Quantity 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.