Learn More
Vitamin K1 2,3-Epoxide (Mixture of Diastereomers), TRC
Product Code. 30005529
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005529 250 mg 250mg
30010598 50 mg 50mg
30007638 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005529 Supplier Toronto Research Chemicals Supplier No. TRCV680013250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25486-55-9
Molecular Formula C31 H46 O3
Molecular Weight (g/mol) 466.7
Synonym Naphth[2,3-b]oxirene-2,7-dione, 1a,7a-dihydro-1a-methyl-7a-(3,7,11,15-tetramethyl-2-hexadecen-1-yl)-, 1,4-Naphthoquinone, 2,3-epoxy-2,3-dihydro-2-methyl-3-phytyl- (6CI,7CI,8CI), Naphth[2,3-b]oxirene-2,7-dione, 1a,7a-dihydro-1a-methyl-7a-(3,7,11,15-tetramethyl-2-hexadecenyl)- (9CI), 1a,7a-Dihydro-1a-methyl-7a-(3,7,11,15-tetramethyl-2-hexadecen-1-yl)naphth[2,3-b]oxirene-2,7-dione, 2,3-Epoxyphylloquinone, Phylloquinone oxide, Phylloquinone, epoxide, Phylloquinone-2,3-epoxide, Vitamin K 2,3-epoxide, Vitamin K epoxide, Vitamin K oxide, Vitamin K1 2,3-epoxide, Vitamin K1 oxide, Vitamin K1, epoxide
IUPAC Name 7a-methyl-1a-[(E)-3,7,11,15-tetramethylhexadec-2-enyl]naphtho[2,3-b]oxirene-2,7-dione
SMILES CC(C)CCCC(C)CCCC(C)CCC\C(=C\CC12OC1(C)C(=O)c3ccccc3C2=O)\C
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C31H46O3/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-31-29(33)27-19-8-7-18-26(27)28(32)30(31,6)34-31/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+
Formula Weight 466.3447
Percent Purity >95%
Chemical Name or Material Vitamin K1 2,3-Epoxide
Grade HPLC