Learn More
Thermo Scientific Chemicals Vitamin B12a hydrochloride
Product Code. 16358297 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16358297
Brand: Thermo Scientific Chemicals 462080050
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58288-50-9
Molecular Formula C62H91ClCoN13O15P
Molecular Weight (g/mol) 1383.84
InChI Key BBQILPBAMXKFOB-UHFFFAOYNA-L
IUPAC Name λ²-cobalt(2+) 4,9,14-tris(2-carbamoylethyl)-3,8,19-tris(carbamoylmethyl)-18-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-2,3,6,8,13,13,16,18-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide hydrate hydrochloride
SMILES O.Cl.[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2[N-]C1=C(C)C1=NC(=CC3=NC(=C(C)C4=NC2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12
Specifications
Quantity 5 g
Formula Weight 1346.35
Chemical Name or Material Vitamin B12a hydrochloride