Learn More
Merck Visualizer Western Blot Detection Kit
Product Code. 10553414 Shop All Merck Products
Click to view available options
Quantity:
20 Blots
Unit Size:
1 set
Product Code. 10553414
Brand: Merck 64202BP
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Detection Method Chemiluminescence
Includes Goat Anti-Rabbit IgG, HRP conjugate, Visualizer™ Detection Reagent B, Visualizer™ Detection Reagent A
Quantity 20 Blots
Packaging 20 Blots
For Use With (Application) For Western Blotting, Dot/Slot Blotting
Host Species Rabbit
Product Type Visualizer Western Blot Detection Kit