Learn More
Vipivotide tetraxetan, MedChemExpress
Product Code. 16494878
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16494878
Brand: Medchem Express HY117410
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Vipivotide tetraxetan (PSMA-617) is a high potent prostate-specific membrane antigen (PSMA) inhibitor, with a Ki of 0.37 nM.
Vipivotide tetraxetan (PSMA-617) is a high potent prostate-specific membrane antigen (PSMA) inhibitor, with a Ki of 0.37 nM.
Specifications
Chemical Name or Material Vipivotide tetraxetan
Physical Form Solid
CAS 1702967-37-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C49H71N9O16
For Use With (Application) Cancer-programmed cell death
Synonym PSMA-617
Solubility Information DMSO : 125 mg/mL (119.95 mM; Need ultrasonic)
SMILES O=C(O)[C@H](CCCCNC([C@H](CC1=CC=C2C=CC=CC2=C1)NC([C@H]3CC[C@H](CNC(CN4CCN(CC(O)=O)CCN(CC(O)=O)CCN(CC(O)=O)CC4)=O)CC3)=O)=O)NC(N[C@H](C(O)=O)CCC(O)=O)=O
Molecular Weight (g/mol) 1042.14
Formula Weight 1042.14
Percent Purity 98.78%
Grade Research