Learn More
Thermo Scientific™ Viper™ and nanoViper™ Fingertight Fittings Accessories
Product Code. p-9495003 Shop All Thermo Scientific Products
Click to view available options
Type:
Filter
Product Code. 15926465
Brand: Thermo Scientific™ 6036.1045
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
Obtain reproducible chromatographic results using tool-free and easy-to-use Thermo Scientific™ Viper™ and Thermo Scientific™ nanoViper™ Fingertight Fittings.
Ensure optimal separations and maximum LC performance using virtually zero-dead-volume connections with the Thermo Scientific™ Viper™ Fingertight Fitting systems. Accessories help complete the installation according to your application needs.
See Viper and nanoViper resources
We offer the following accessories for Viper Fingertight Fittings Systems:
  • Union exclusively designed for connecting Viper and nanoViper Fingertight Fittings directly without additional dead volume
  • Viper Plug for installation in open valve positions to protect the bores or for diagnostic functions
  • Tool to recalibrate Viper and nanoViper fittings for an extended lifetime when connected to multiple columns with non-standard column bore geometry
  • Viper Inline Filter (titanium, av. frit pre size 0.5μm) to protect the column from contaminants
  • Adaptor for Viper Fingertight Fittings (2 pcs, stainless steel, for columns ≥ 2.1 mm ID) to adapt to non-standard column bore geometry
Warranty
Viper and nanoViper Fingertight Fittings Accessories
Specifications
Description Viper Inline Filter
For Use With (Application) Column Protection
For Use With (Equipment) Analytical HPLC and UHPLC Systems
Material Titanium
Max. Pressure 1500 bar
Type Filter
Product Line Viper