missing translation for 'onlineSavingsMsg'
Learn More

Vinexin Antibody, Novus Biologicals™

Product Code. 18404041 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404041 25 μL 25µL
18462181 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18404041 Supplier Novus Biologicals Supplier No. NBP18708425ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Vinexin Polyclonal antibody specifically detects Vinexin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vinexin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias SCAM-1SH3-containing adapter molecule 1, SCAM1vinexin beta (SH3-containing adaptor molecule-1), SH3D4VINEXIN, sorbin and SH3 domain containing 3, Sorbin and SH3 domain-containing protein 3, vinexin
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: GPASAWSSSYPHAPYLGSARSLSPHKMADGGSPFLGRRDFVYPSSTRDPSASNGGGSPARREEKKRKAARLKFDFQAQSPKELTLQKGDIVYIHKEVDKNWL
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10174
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.