Learn More
MedChemExpress Vincristine sulfate
Product Code. 16458998
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16458998 Supplier MedChemExpress Supplier No. HYN0488
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
MedChemExpress Vincristine sulfate is an antitumor vinca alkaloid which inhibits microtubule formation in mitotic spindle, resulting in an arrest of dividing cells at the metaphase stage. It binds to microtubule with a Ki of 85 nM.
Specifications
Content And Storage 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Molecular Weight (g/mol) 923.04 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 2068-78-2
Product Type Natural Products
Purity 97.83%
Solubility DMSO : 83.33 mg/mL (90.28 mM∣ultrasonic and warming and heat to 80°C) ∣H2O : 50 mg/mL (54.17 mM∣Need ultrasonic)
Synonym Leurocristine sulfate∣NSC-67574 sulfate∣22-Oxovincaleukoblastine sulfate
Target Microtubule/Tubulin∣Apoptosis
Quantity 10 mg