Learn More
Vildagliptin, TRC
Product Code. 30008315
Change view
Click to view available options
Quantity:
10 mg
500 mg
1 g
Unit Size:
10mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008315 1 g 1g
17931855 10 mg 10mg
17951535 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008315 Supplier Toronto Research Chemicals Supplier No. TRCV3050001G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 274901-16-5
Molecular Formula C17 H25 N3 O2
Molecular Weight (g/mol) 303.4
Synonym (2S)-1-[2-[(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)amino]acetyl]-2-pyrrolidinecarbonitrile, 2-Pyrrolidinecarbonitrile, 1-[[(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)amino]acetyl]-, (2S)- (9CI), (S)-1-[2-[(3-Hydroxyadamant-1-yl)amino]acetyl]pyrrolidine-2-carbonitrile, DSP 7238, Equa, Galvus, LAF 237, NVP-LAF 237, Vildagliptin
IUPAC Name (2S)-1-[2-[(3-hydroxy-1-adamantyl)amino]acetyl]pyrrolidine-2-carbonitrile
SMILES OC12CC3CC(C1)CC(C3)(C2)NCC(=O)N4CCC[C@H]4C#N
Specifications
Quantity 1 g
InChI Formula InChI=1S/C17H25N3O2/c18-9-14-2-1-3-20(14)15(21)10-19-16-5-12-4-13(6-16)8-17(22,7-12)11-16/h12-14,19,22H,1-8,10-11H2/t12?,13?,14-,16?,17?/m0/s1
Formula Weight 303.1947
Percent Purity >95%
Chemical Name or Material Vildagliptin
Grade HPLC