Learn More
Ethyl-alpha,alpha-d2-benzene-d5, CDN
Specifications
Specifications
| Chemical Name or Material | Ethylbenzene-alpha,alpha,2,3,4,5,6-D7 |
| CAS | 84272-90-2 |
| Molecular Formula | C8 D7 H3 |
| Quantity | 0.1 g |
| Synonym | Benzene-d5, ethyl-1,1-d2- (9 CI, ACI), Ethyl-1,1-d2benzene-d5 (ACI), 1,2,3,4,5-Pentadeuterio-6-(1,1-dideuterioethyl)benzene, Ethyl-α,α-d2-benzene-d5, Pentadeutero(1,1-dideuteroethyl)benzene, Ethylbenzene-d7, Ethylbenzene-α,α,2,3,4,5,6-d7, Ethylbenzene-α,α,2,3,4,5,6-D7 |
| InChI Formula | InChI=1 S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i2D2,3 D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(1,1-dideuterioethyl)benzene |
| Molecular Weight (g/mol) | 113.2081 |
| Formula Weight | 113.1222 g/mol |
| Show More |