Learn More
2,2',4,6,6'-Pentachlorobiphenyl-3',4',5'-d3, CDN
Specifications
Specifications
| Chemical Name or Material | 2,2',4,6,6'-Pentachlorobiphenyl-3',4',5'-D3 |
| CAS | 1219794-64-5 |
| Molecular Formula | C12 D3 H2 Cl5 |
| Quantity | 0.01 g |
| Synonym | 2,2',4,6,6'-Pentachloro-1,1'-biphenyl-3',4',5'-d3, 2,2',4,6,6'-Pentachlorobiphenyl-3',4',5'-D3, 2,2',4',6,6'-Pentachlorobiphenyl-3,4,5-d3, PCB 104-d3, 2,2',4,6,6'-PentaCB-3',4',5'-d3 |
| InChI Formula | InChI=1 S/C12H5Cl5/c13-6-4-9(16)12(10(17)5-6)11-7(14)2-1-3-8(11)15/h1-5 H/i1D,2 D,3 D |
| SMILES | [2 H]c1c([2 H])c(Cl)c(c(Cl)c1[2 H])c2c(Cl)cc(Cl)cc2Cl |
| IUPAC Name | 1,5-dichloro-2,3,4-trideuterio-6-(2,4,6-trichlorophenyl)benzene |
| Molecular Weight (g/mol) | 329.4516 |
| Formula Weight | 326.9022 g/mol |
| Show More |