Learn More
Invitrogen™ Human MESDC1 (aa 301-362) Control Fragment Recombinant Protein
Product Code. 30208418
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy
Product Code. 30208418
Brand: Invitrogen™ RP108587
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84871 (PA5-84871. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Actin-binding protein which may have an oncogenic function and regulates cell proliferation, migration and invasion in cancer cells.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q9H1K6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 59274
Name Human MESDC1 (aa 301-362) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW061151; MESDC1; Mesoderm development candidate 1; mesoderm development candidate 1; talin rod domain containing 1; talin rod domain containing 1; talin rod domain-containing protein 1; TLNRD1
Common Name MESDC1
Gene Symbol Tlnrd1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLTQCLRDLAQHPDGGAKMSDHRERLRNSACAVSEGCTLLSQALRERSSPRTLPPVNSNSVN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining