Learn More
Invitrogen™ Human ESPN (aa 785-848) Control Fragment Recombinant Protein
Product Code. 30202511
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy
Product Code. 30202511
Brand: Invitrogen™ RP94521
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55941 (PA5-55941. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Multifunctional actin-bundling protein. Plays a major role in regulating the organization, dimensions, dynamics and signaling capacities of the actin filament-rich, microvillus-type specializations that mediate sensory transduction in variouS mechanosensory and chemosensory cells.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number B1AK53
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 83715
Name Human ESPN (aa 785-848) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias actin cytoskeletal regulatory protein; autosomal recessive deafness type 36 protein; DFNB36; Ectoplasmic specialization protein; espin; Espn; je; jerker; Jerker, deafness locus; LP2654
Common Name ESPN
Gene Symbol Espn
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SMPAWRRDLLRKKLEEEREQKRKEEERQKQEELRREKEQSEKLRTLGYDESKLAPWQRQVILKK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining