Learn More
CD7 Antibody (grisnilimAb) [Janelia Fluor™ 525], Novus Biologicals™
Product Code. 30162394
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30162394 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30162394 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328084JF525
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Human Monoclonal Antibody
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Antigen CD7
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone grisnilimAb
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
Host Species Human
Immunogen CD7
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 924
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2