Learn More
Human/Mouse PRDM16/MEL1 Antibody, R&D Systems™
Description
PRDM16/MEL1 Polyclonal specifically detects PRDM16/MEL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antigen | PRDM16/MEL1 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | A2A935.1 |
| Gene Alias | MDS1/EVI1-like gene 1, MEL1, MEL1KIAA1675PFM13MGC166915, PFM13, PR domain containing 16, PR domain zinc finger protein 16, PR domain-containing protein 16, Transcription factor MEL1 |
| Gene Symbols | PRDM16 |
| Show More |
For Research Use Only