Learn More
Human Tsukushi/TSK Antibody, R&D Systems™
Description
Tsukushi/TSK Polyclonal specifically detects Tsukushi/TSK in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
| Antigen | Tsukushi/TSK |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 5-15 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q8WUA8 |
| Gene Alias | E2IG4, E2IG4E2-induced gene 4 protein, leucine rich repeat containing 54, Leucine-rich repeat-containing protein 54, LRRC54, TSKtsukushi homolog, TSKU, Tsukushi, tsukushi small leucine rich proteoglycan homolog (Xenopus laevis), tsukushi, small leucine rich proteoglycan, tsukushin |
| Gene Symbols | TSKU |
| Show More |
For Research Use Only